missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP2B4, Mouse anti-Human, Clone: 2G8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00000493-M07.100ug
This item is not returnable.
View return policy
Description
Sequence: MTNPSDRVLPANSMAESREGDFGCTVMELRKLMELRSRDALTQINVHYGGVQNLCSRLKTSPVEGLSGNPADLEKRRQVFGHNVIPPKKPKTSpecifications
| ATP2B4 | |
| Monoclonal | |
| Unconjugated | |
| ATPase, Ca++ transporting, plasma membrane 4 | |
| ATP2B2/DKFZp686G08106/DKFZp686M088/MXRA1/PMCA4/PMCA4b/PMCA4x | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2G8 | |
| In 1x PBS, pH 7.4 | |
| NM_001684 | |
| ATP2B4 | |
| ATP2B4 (NP_001675, 1 a.a. ∼ 92 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 493 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction