Learn More
Invitrogen™ ATP4B Monoclonal Antibody (2G11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA3923
Description
MA3-923 detects the beta-subunit of hydrogen/potassium ATPase from bovine, human, canine, porcine, rabbit, mouse, ferret, and rat tissues. MA3-923 has been successfully used in Western blot, immunohistochemistry, immunocytochemistry, and immunoprecipitation procedures (ChIP). By Western blot, this antibody detects various forms of the beta-subunit, including a 60 to 80 kDa glycosylated protein, a 52 kDa beta-subunit precursor, and the 34 kDa core peptide from rabbit stomach extracts. Immunohistochemical staining of hydrogen/potassium ATPase in rabbit gastric glands with MA3-923 results in specific staining of parietal cells. MA3-923 has been shown to inhibit the enzymatic activity of the hydrogen/potassium ATPase and to alter the affinity of the cytoplasmic potassium binding site. The MA3-923 antigen is purified 34 kDa core peptide from deglycosylated hog gastric microsomes. This antibody recognizes an epitope between amino acid residues 1-13 or 15-28 located on the cytoplasmic side of the beta-subunit.
The hydrogen/potassium ATPase, or gastric proton pump, belongs to a family of P-type cation-transporting ATPases. This family of ATPases shares a number of functional and structural similarities including the common feature of consisting of an alpha and beta subunit. Like the ubiquitous sodium/potassium ATPase, the hydrogen/potassium ATPase consists of a large transmembrane catalytic subunit, termed the alpha- subunit which contains sites for ATP binding and phosphorylation, and an associated smaller glycoprotein, termed the beta-subunit which may play a role in maintaining the structural and functional integrity of the complex.
Specifications
| ATP4B | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites, PBS with 0.05% sodium azide | |
| P18434, P18597, P18598, P33704, P50992, P51164 | |
| ATP4B | |
| Purified 34 kDa core peptide from deglycosylated hog gastric microsomes. | |
| RUO | |
| 100009125, 11945, 24217, 397131, 404020, 496, 614308 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 2G11 | |
| Unconjugated | |
| ATP4B | |
| (H+,K+)-ATPase beta-subunit; ATP4B; ATP6B; ATPase H+/K+ transporting beta subunit; ATPase H+/K+ transporting subunit beta; ATPase H+K+-transporting beta / (gastric HK-ATPase beta subunit) defined by SSR; ATPase, H+,K+-transporting, beta / (gastric H,K-ATPase beta subunit), defined by SSR; ATPase, H+/K+ exchanging, beta polypeptide; ATPase, H+/K+ transporting, beta polypeptide; ATPase, H+/K+ transporting, beta polypeptide, gastric specific; AV080843; gastric H(+)/K(+) ATPase subunit beta; gastric H+/K+ ATPase beta subunit; gastric H+/K+-ATPase beta subunit; gastric hydrogen-potassium ATPase, beta; gp60-90; H,K-ATPase-Beta; H.K-ATPase, beta subunit; H+,K+-ATPase; H+/K+ ATPase beta; H+/K+ ATPase beta subunit; H+/K+-ATPase beta; H+/K+-ATPase beta subunit; potassium-transporting ATPase beta chain; potassium-transporting ATPase subunit beta; Proton pump beta chain; S76401S1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Bovine, Pig, Mustelid | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.