missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP5A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12789-50ul
This item is not returnable.
View return policy
Description
ATP5A Polyclonal antibody specifically detects ATP5A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ATP5A | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform1, cardiac muscle, ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform2, non-cardiac muscle-like 2, ATP sythase (F1-ATPase) alpha subunit, ATP5AL2, ATP5AMOM2, ATPMATP synthase subunit alpha, mitochondrial, cardiac muscle, EC 3.6.3, EC 3.6.3.14, hATP1, mitochondrial ATP synthetase, oligomycin-resistant, OMR, ORMATP synthase alpha chain, mitochondrial | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ATP5A | |
| 50 μg | |
| Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 498 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction