Learn More
ATP5C1 Mouse anti-Human, CoraLite 594, Clone: 1B5A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60284-100UL
Description
ATP5C1(ATP synthase subunit gamma, mitochondrial) is also named as ATP5C, ATP5CL1 and belongs to the ATPase gamma chain family. Some scientists reported the complete sequence of the gene for the human ATP synthase gamma subunit and described tissue-specific isoforms of the subunit generated by alternative splicing of exon 9. The liver (L) isoform differed from the heart (H) isoform by the addition of a single amino acid (asp273) at the C terminus. (PMID:8227057).
Specifications
| ATP5C1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| IgG2a |
| Immunofluorescence | |
| 1B5A2 | |
| CoraLite 594 | |
| 100 μL | |
| 509 | |
| Liquid |