Learn More
ATP5C1 Mouse anti-Human, CoraLite 594, Clone: 1B5A2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-60284-100UL
Product Code. 17214796
  • € 450.00 / 100µL
Description
ATP5C1(ATP synthase subunit gamma, mitochondrial) is also named as ATP5C, ATP5CL1 and belongs to the ATPase gamma chain family. Some scientists reported the complete sequence of the gene for the human ATP synthase gamma subunit and described tissue-specific isoforms of the subunit generated by alternative splicing of exon 9. The liver (L) isoform differed from the heart (H) isoform by the addition of a single amino acid (asp273) at the C terminus. (PMID:8227057).
Specifications
Specifications
ATP5C1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
IgG2a
Immunofluorescence
1B5A2
CoraLite 594
100 μL
509