Learn More
ATP5F1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-15999-20UL
 View more versions of this product
Product Code. 18075155
  • € 145.00 / 20µL
Description
The ATP5F1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ATP5F1. This antibody recognizes human, mouse antigen. The ATP5F1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
ATP5F1
1000 μg/mL
ATP5F1, ATPase subunit b, PIG47
20 μL
515
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human, Mouse
150 kDa