missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP5G1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA575872

Product Code. 15982395

  • € 526.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Mitochondrial ATP synthase is composed of two multi-subunit complexes that utilize an inner membrane electrochemical gradient to catalyze the synthesis of ATP during oxidative phosphorylation. The two multi-subunit complexes are designated F1 and F0, the former of which comprises the soluble catalytic core and the latter of which comprises the membrane-spanning proton channel of ATP synthase. F1 consists of five distinct subunits, designated ATP5A, ATP5B, ATP5C1, ATP5D and ATP5E, while F0 consists of ten subunits, designated ATP5H, ATP5G1, ATP5I, ATP5G2, ATP5J2, ATP5J, ATP5G3, ATP5S, ATP5F1 and ATP5L. ATP5G1, ATP5G2 and ATP5G3 localize to the mitochondrial membrane and exist as subunits of the F0 complex. ATP5G2 exists as three alternatively spliced isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATP5G1
Polyclonal
Unconjugated
ATP5MC1
ATP Synthase C; ATP synthase F(0) complex subunit C1, mitochondrial; ATP synthase H+ transporting mitochondrial F0 complex subunit c (subunit 9) isoform 1; ATP synthase lipid-binding protein; ATP synthase lipid-binding protein, mitochondrial; ATP synthase membrane subunit c locus 1; ATP synthase proteolipid P1; ATP synthase proton-transporting mitochondrial F(0) complex subunit C1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9), isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c, isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C1 (subunit 9); ATP5A; ATP5G; ATP5G1; ATP5MC1; ATPase protein 9; ATPase subunit 9; ATPase subunit c; mitochondrial ATP synthase, subunit 9, isoform 1; mitochondrial ATP synthase, subunit C, isoform 1
Rabbit
Antigen Affinity Chromatography
RUO
11951, 29754, 516
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P05496, Q06645, Q9CR84
ATP5MC1
Synthetic peptide corresponding to residues in Human ATP Synthase C.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.