Learn More
Novus Biologicals™ ATP5H Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30298
Product Code. 30047323
  • € 571.00 / 100µg
Description
ATP5H Polyclonal antibody specifically detects ATP5H in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATP5H
Polyclonal
Tris-citrate/phosphate pH 7 -8.
Rabbit
Affinity purified
RUO
Primary
Human, Mouse
Purified
Immunoprecipitation, Western Blot
Unconjugated
ATP synthase D chain, mitochondrial, ATP synthase subunit d, mitochondrial, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit d, ATP synthase, H+ transporting, mitochondrial F1F0, subunit d, ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d, ATP5JD, ATPase subunit d, ATPQ, My032 protein
Between 111 and 161
100 μg
Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
10476
Store at 4°C. Do not freeze.