Learn More
ATP5J2 Mouse, anti-Human, CoraLite Plus 488, Clone: 2B3C3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-68128-100UL
 View more versions of this product
Product Code. 18088735
  • € 450.00 / 100µL
Description
The ATP5J2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATP5J2. This antibody recognizes human, mouse, rat, rabbit antigen. The ATP5J2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
ATP5J2
2B3C3
CoraLite Plus 488
Mouse
Primary
Human, Mouse, Rat, Rabbit
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
ATP5J2, ATP5JL
100 μL
9551
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.