missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP5O Antibody (OTI2E9), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70239AF488
This item is not returnable.
View return policy
Description
ATP5O Monoclonal specifically detects ATP5O in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ATP5O | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ATP5PO | |
| Recombinant protein fragment corresponding to amino acids 24-213 of human ATP5O (NP_001688) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2E9 | |
| Western Blot, Immunohistochemistry | |
| ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit, human ATP synthase OSCP subunit, oligomycin sensitivity conferring protein10ATPOATP synthase subunit O, mitochondrial, Oligomycin sensitivity conferral protein, oligomycin sensitivity conferring protein, OSCPhuman ATP synthase OSCP subunit | |
| Mouse | |
| Affinity Purified | |
| Core ESC Like Genes, Stem Cell Markers | |
| 539.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction