missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP5SL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546192
This item is not returnable.
View return policy
Description
Peptide sequence: ISDLPAVSNP GLTQILVEEM LPNCEVVGVD WAEGLKSGPE EQPRDTASPV Sequence homology: Guinea Pig: 77%; Human: 100%; Rabbit: 91%.
Required for the assembly of the mitochondrial NADH:ubiquinone oxidoreductase complex (complex I). Involved in the assembly of the distal region of complex I. [UniProt].
Specifications
| ATP5SL | |
| Polyclonal | |
| Unconjugated | |
| Dmac2 | |
| ATP synthase subunit s-like protein; ATP5S like; Atp5sl; ATP5S-like; distal membrane arm assembly complex 2; distal membrane-arm assembly complex protein 2; DMAC2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55101 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NW81 | |
| Dmac2 | |
| Synthetic peptide directed towards the C-terminal of human ATP5SL (aa 206-255). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction