missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V0D1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99561C
This item is not returnable.
View return policy
Description
ATP6V0D1 Polyclonal antibody specifically detects ATP6V0D1 in Human samples. It is validated for Western Blot
Specifications
| ATP6V0D1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 9114 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 32 kDa accessory protein, ATP6D, ATP6DV, ATP6V0, ATPase, H+ transporting, lysosomal (vacuolar proton pump), member D, ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d isoform 1, ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d1, FLJ43534, H(+)-transporting two-sector ATPase, subunit D, P39, Vacuolar proton pump subunit d 1, V-ATPase 40 kDa accessory protein, V-ATPase AC39 subunit, V-ATPase, subunit D, VATX, Vma6, VPATPDV-ATPase subunit d 1, V-type proton ATPase subunit d 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ATP6V0D1. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction