missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP6V0D2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544359
This item is not returnable.
View return policy
Description
Peptide sequence: GLRLLAQAED FDQMKNVADH YGVYKPLFEA VGGSGGKTLE DVFYEREVQM Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
ATP6V0D2 is the subunit of the integral membrane V0 complex of vacuolar ATPase. Vacuolar ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells, thus providing most of the energy required for transport processes in the vacuolar system. ATP6V0D2 may play a role in coupling of proton transport and ATP hydrolysis.
Specifications
| ATP6V0D2 | |
| Polyclonal | |
| Unconjugated | |
| ATP6V0D2 | |
| 1620401A02Rik; AI324824; ATP6D2; Atp6v0d2; ATPase H+ transporting V0 subunit d2; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d2; ATPase, H+ transporting, lysosomal V0 subunit D2; ATPase, H+ transporting, V0 subunit D, isoform 2; FLJ38708; osteoclast-specific vacuolar ATP synthase; similar to ATPase, H+ transporting, V0 subunit D isoform 2; vacuolar proton pump subunit d 2; vacuolar proton-translocating ATPase d subunit d2; V-ATPase; V-ATPase subunit d 2; VMA6; V-type proton ATPase subunit d 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 245972 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N8Y2 | |
| ATP6V0D2 | |
| Synthetic peptide directed towards the middle region of human ATP6V0D2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction