missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ATP6V1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29501
This item is not returnable.
View return policy
Description
ATP6V1A Polyclonal antibody specifically detects ATP6V1A in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| ATP6V1A | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 523 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| 70kD, isoform 1, ATP6A1, ATP6V1A1ATPase, H+ transporting, lysosomal, subunit A1, ATPase, H+ transporting, lysosomal (vacuolar proton pump), alpha polypeptide, ATPase, H+ transporting, lysosomal 70kDa, V1 subunit A, EC 3.6.3, EC 3.6.3.14, H(+)-transporting two-sector ATPase, subunit A, H+-transporting ATPase chain A, vacuolar (VA68 type), HO68, VA68, vacuolar ATP synthase catalytic subunit A, ubiquitous isoform, Vacuolar ATPase isoform VA68, vacuolar proton pump alpha subunit 1, Vacuolar proton pump subunit alpha, V-ATPase 69 kDa subunit, V-ATPase 69 kDa subunit 1, V-ATPase A subunit 1, V-ATPase subunit A, Vma1, VPP2, V-type proton ATPase catalytic subunit A | |
| Between 567 and 617 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction