missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1B1 Antibody (OTI1H6), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70237AF405
This item is not returnable.
View return policy
Description
ATP6V1B1 Monoclonal specifically detects ATP6V1B1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ATP6V1B1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ATP6V1B1 | |
| Full length recombinant protein of human ATP6V1B1 (NP_001683) produced in HEK293T cells. | |
| 0.1 mL | |
| 525.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1H6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ATP6B158kD subunit, ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1, Endomembrane proton pump 58 kDa subunit, H+-ATPase beta 1 subunit, vacuolar proton pump 3, Vacuolar proton pump subunit B 1, vacuolar proton pump, subunit 3, VATBMGC32642, V-ATPase B1 subunit, V-ATPase subunit B 1, Vma2, V-type proton ATPase subunit B, kidney isoform | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction