missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1B2 Antibody (OTI1E11), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70238H
This item is not returnable.
View return policy
Description
ATP6V1B2 Monoclonal specifically detects ATP6V1B2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ATP6V1B2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| ATP6V1B2 | |
| Full length recombinant protein of human ATP6V1B2 (NP_001684) produced in HEK293T cells. | |
| 0.1 mL | |
| 526.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1E11 | |
| Western Blot, Immunohistochemistry | |
| 000 subunit, 56/58kD, isoform 2, ATP6B2ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta polypeptide, ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2, Endomembrane proton pump 58 kDa subunit, H+ transporting two-sector ATPase, HO57ATP6B1B2, vacuolar H+-ATPase 56, Vacuolar proton pump subunit B 2, VATB, V-ATPase B2 subunit, V-ATPase subunit B 2, Vma2, VPP3ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B, isoform 2, V-type proton ATPase subunit B, brain isoform | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction