missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p107 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97802PCP
This item is not returnable.
View return policy
Description
p107 Polyclonal antibody specifically detects p107 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| p107 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| cellular protein 107,107 kDa retinoblastoma-associated protein, cp107, p107MGC40006, PRB1, retinoblastoma-like 1 (p107), retinoblastoma-like protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human p107 fragment. | |
| 0.1 mL | |
| Cancer | |
| 5933 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction