missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1C2 Mouse anti-Human, Clone: OTI3E9, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808257S
This item is not returnable.
View return policy
Description
ATP6V1C2 Monoclonal antibody specifically detects ATP6V1C2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ATP6V1C2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NEY4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 245973 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3E9 | |
| Unconjugated | |
| Atp6v1c2 | |
| 1110038G14Rik; Atp6c2; Atp6v1c2; ATPase H+ transporting V1 subunit C2; ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C2; ATPase, H+ transporting, lysosomal V1 subunit C2; ATPase, H+ transporting, V1 subunit C, isoform 2; vacuolar H+ ATPase C2; vacuolar proton pump subunit C 2; V-ATPase C2 subunit; V-ATPase subunit C 2; VMA5; V-type ATPase C2 subunit a isoform; V-type proton ATPase subunit C 2 | |
| Full length human recombinant protein of human ATP6V1C2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction