missing translation for 'onlineSavingsMsg'
Learn More

ATP6V1D Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. p-200061699 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18694852 0.02 mL 0.02mL
18641042 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18694852 Supplier Novus Biologicals Supplier No. NBP2927090.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP6V1D Polyclonal antibody specifically detects ATP6V1D in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V1D
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ATP6M, ATPase, H+ transporting lysosomal, member M, ATPase, H+ transporting, lysosomal (vacuolar proton pump), ATPase, H+ transporting, lysosomal 34kDa, V1 subunit D, H(+)-transporting two-sector ATPase, subunit M, vacuolar ATP synthase subunit D, vacuolar H-ATPase subunit D, vacuolar proton pump D subunit, vacuolar proton pump delta polypeptide, Vacuolar proton pump subunit D, vacuolar proton-ATPase subunit D, VATDV-ATPase D subunit, V-ATPase 28 kDa accessory protein, V-ATPase subunit D, VMA8, V-type proton ATPase subunit D
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-247 of human ATP6V1D (NP_057078.1). MSGKDRIEIFPSRMAQTIMKARLKGAQTGRNLLKKKSDALTLRFRQILKKIIETKMLMGEVMREAAFSLAEAKFTAGDFSTTVIQNVNKAQVKIRAKKDNVAGVTLPVFEHYHEGTDSYELTGLARGGEQLAKLKRNYAKAVELLVELASLQTSFVTLDEAIKITNRRVNAIEHVIIPRIERTLAYIITELDEREREEFYRLKKIQEKKKILKEKSEKDLEQRRAAGEVLEPANLLAEEKDEDLLFE
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51382
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.