missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1F Monoclonal Antibody (OTI6C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502322
This item is not returnable.
View return policy
Description
ATP6V1F encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c′, c", and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is the V1 domain F subunit protein.Specifications
| ATP6V1F | |
| Monoclonal | |
| 0.44 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P50408, Q16864 | |
| Atp6v1f | |
| Full length human recombit protein of human ATP6V1F produced in E.coli. | |
| 100 μL | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI6C11 | |
| Unconjugated | |
| ATP6V1F | |
| ATP6S14, VATF, Vma7 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 116664, 475199, 9296 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction