missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1F Monoclonal Antibody (OTI6C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502322S
This item is not returnable.
View return policy
Description
ATP6V1F Monoclonal antibody specifically detects ATP6V1F in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| ATP6V1F | |
| Monoclonal | |
| 0.44 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00408 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5138 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI6C11 | |
| Unconjugated | |
| PDE2A | |
| 1110004G16Rik; adenosinetriphosphatase 14k chain; ATP6S14; ATP6V1F; ATPase H+ transporting V1 subunit F; ATPase, H transporting, lysosomal V1 subunit F; ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F; ATPase, H+ transporting, lysosomal V1 subunit F; ATPase, H+ transporting, V1 subunit F; ATPase, vacuolar, 14 kD; H(+)-transporting two-sector ATPase, 14kD subunit; vacuolar ATP synthase subunit F; vacuolar proton pump F subunit; vacuolar proton pump subunit F; VATF; V-ATPase 14 kDa subunit; V-ATPase F subunit; V-ATPase subunit F; Vma7; V-type proton ATPase subunit F; v-type proton ATPase subunit F-like | |
| Full length human recombinant protein of human PDE2A produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction