missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1G3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88894-25ul
This item is not returnable.
View return policy
Description
ATP6V1G3 Polyclonal specifically detects ATP6V1G3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ATP6V1G3 | |
| Polyclonal | |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000-1:2500 | |
| ATPase, H+ transporting, lysosomal 13kD, V1 subunit G isoform 3, ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G isoform 3, ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G3, H+ transporting, lysosomal (vacuolar proton pump) subunit G3, MGC119810, MGC119813, vacuolar ATP synthase subunit G 3, vacuolar proton pump G subunit 3, Vacuolar proton pump subunit G 3, vacuolar proton pump, subunit G3, V-ATPase 13 kDa subunit 3, V-ATPase G subunit 3, V-ATPase G3 subunit, V-ATPase subunit G 3, V-type proton ATPase subunit G 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 127124 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ATP6V1G3 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AKEEAMVEIDQYRMQRDKEFRLKQSKIMGSQNNLSDEIEEQTLGKIQELNGHYNKYMESVMNQLLSMVCDMKPEIHVNYR | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction