missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP7b Antibody (9H8), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | ATP7b |
|---|---|
| Clone | 9H8 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Flow Cytometry, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30025117
|
Novus Biologicals
NBP3-26489-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30024227
|
Novus Biologicals
NBP3-26489-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATP7b Monoclonal antibody specifically detects ATP7b in Human samples. It is validated for Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
| ATP7b | |
| Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ATPase, Cu(2+)- transporting, beta polypeptide, ATPase, Cu++ transporting, beta polypeptide, Copper pump 2, copper-transporting ATPase 2, EC 3.6.3, EC 3.6.3.4, PWD, WC1, WD, Wilson disease-associated protein, WNDATPase, Cu++ transporting, beta polypeptide (Wilson disease) | |
| Recombinant Human ATP7b protein [UniProt P35670] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 9H8 | |
| Flow Cytometry, ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 540 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title