missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP7b Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-361AF532
This item is not returnable.
View return policy
Description
ATP7b Polyclonal antibody specifically detects ATP7b in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| ATP7b | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| ATPase, Cu(2+)- transporting, beta polypeptide, ATPase, Cu++ transporting, beta polypeptide, Copper pump 2, copper-transporting ATPase 2, EC 3.6.3, EC 3.6.3.4, PWD, WC1, WD, Wilson disease-associated protein, WNDATPase, Cu++ transporting, beta polypeptide (Wilson disease) | |
| A synthetic peptide made to an internal sequence near the C-terminus of human ATP7b. [UniProt# P35670] | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 540 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat, Mouse (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction