Learn More
Invitrogen™ ATP7B Polyclonal Antibody, Biotin
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA522767
Product Code. 13257208
  • € 526.00 / 100µL
Description
This antibody does not react with mouse samples.
This gene is a member of the P-type cation transport ATPase family and encodes a protein with several membrane-spanning domains, an ATPase consensus sequence, a hinge domain, a phosphorylation site, and at least 2 putative copper-binding sites. This protein functions as a monomer, exporting copper out of the cells, such as the efflux of hepatic copper into the bile. Alternate transcriptional splice variants, encoding different isoforms with distinct cellular localizations, have been characterized. Mutations in this gene have been associated with Wilson disease (WD).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATP7B
Polyclonal
Biotin
Atp7b
Atp7a; ATP7B; ATPase copper transporting beta; ATPase, Cu(2+)- transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide (same as Wilson disease); copper pump 2; Copper-transporting ATPase 2; Hts; PINA; PINA gene, promoter; pineal night-specific ATPase; PWD; RP11-327P2.1; RP11-327P2.3; toxic milk; tx; WC1; WD; Wilson disease-associated protein; wilson disease-associated protein homolog; Wilson protein; Wnd; WND/140 kDa
Rabbit
Antigen affinity chromatography
RUO
11979, 24218, 540
4°C, store in dark
Liquid
Western Blot, Immunocytochemistry
1.01 mg/mL
PBS with 0.05% sodium azide
P35670, Q64446, Q64535
Atp7b
A synthetic peptide made to an internal sequence near the C-terminus of human ATP7b.
100 μL
Primary
Human, Mouse, Rat