missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP7B Polyclonal Antibody
GREENER_CHOICE

Product Code. 17762334
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17762334 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17762334 Supplier Invitrogen™ Supplier No. PA5120938

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: BT-474, HL-60, Mouse liver. The target is usually found in the following locations: Cytoplasm, Golgi apparatus, Golgi apparatus membrane, Mitochondrion, Multi-pass membrane protein, Multi-pass membrane protein, trans-Golgi network membrane. Immunogen sequence: SSVSVVLSSL QLKCYKKPDL ERYEAQAHGH MKPLTASQVS VHIGMDDRWR DSPRATPWDQ VSYVSQVSLS SLTSDKPSRH SAAADDDGDK WSLLLNGRDE E.

This gene is a member of the P-type cation transport ATPase family and encodes a protein with several membrane-spanning domains, an ATPase consensus sequence, a hinge domain, a phosphorylation site, and at least 2 putative copper-binding sites. This protein functions as a monomer, exporting copper out of the cells, such as the efflux of hepatic copper into the bile. Alternate transcriptional splice variants, encoding different isoforms with distinct cellular localizations, have been characterized. Mutations in this gene have been associated with Wilson disease (WD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP7B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Atp7b
Gene Accession No. P35670, Q64535
Gene Alias Atp7a; ATP7B; ATPase copper transporting beta; ATPase, Cu(2+)- transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide (same as Wilson disease); copper pump 2; Copper-transporting ATPase 2; Hts; PINA; PINA gene, promoter; pineal night-specific ATPase; PWD; RP11-327P2.1; RP11-327P2.3; toxic milk; tx; WC1; WD; Wilson disease-associated protein; wilson disease-associated protein homolog; Wilson protein; Wnd; WND/140 kDa
Gene Symbols Atp7b
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1251-1351 of human ATP7B (NP_0012301111).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24218, 540
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.