missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488 Anti-Human CD62L (DREG56), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-65167-100TESTS
This item is not returnable.
View return policy
Description
The CD62L antibody from Proteintech is a mouse monoclonal antibody generated with pma-activated human peripheral blood leukocytes of human CD62L. This antibody recognizes human antigen. The CD62L antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 488.Specifications
| CD62L | |
| Monoclonal | |
| 5 μL/test | |
| CD62L; LAM-1; LAM1; LECAM1; LNHR; LSEL; LYAM1; Leu-8; Lyam-1; PLNHR; TQ1; hLHRc | |
| PMA-activated human peripheral blood leukocytes | |
| 100 Tests | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| DREG56 | |
| CoraLite Plus 488 | |
| Mouse | |
| Purified by protein-A affinity chromatography | |
| RUO | |
| 6402 | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction