missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP8B3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17781984
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17781984 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17781984 Supplier Invitrogen™ Supplier No. PA5120588

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HCT116.

P4-ATPase flippase which catalyzes the hydrolysis of ATP coupled to the transport of aminophospholipids from the outer to the inner leaflet of various membranes and ensures the maintenance of asymmetric distribution of phospholipids. Phospholipid translocation seems also to be implicated in vesicle formation and in uptake of lipid signaling molecules. May be responsible for the maintenance of asymmetric distribution of phosphatidylserine (PS) in spermatozoa membranes. Involved in acrosome reactions and binding of spermatozoa to zona pellucida.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP8B3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ATP8B3
Gene Accession No. O60423
Gene Alias aminophospholipid translocase ATP8B3; ATP1K; ATP8B3; ATPase class I type 8B member 3; ATPase phospholipid transporting 8B3; ATPase, aminophospholipid transporter, class I, type 8B, member 3; ATPase, class I, type 8B, member 3; ATPIK; FOS37502_2; Phospholipid-transporting ATPase IK; potential phospholipid-transporting ATPase IK; probable phospholipid-transporting ATPase IK
Gene Symbols ATP8B3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148229
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.