missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATPAF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565022
This item is not returnable.
View return policy
Description
Immunogen sequence: KFDLIWNRAQS CPTFLCALPR REGYEFFVGQ WTGTELHFTA LINIQTRGEA AASQLILYHY PELKEE Highest antigen sequence identity to the following orthologs - mouse 97%, rat 96%.
This gene encodes an assembly factor for the F(1) component of the mitochondrial ATP synthase. This protein binds specifically to the F1 beta subunit and is thought to prevent this subunit from forming nonproductive homooligomers during enzyme assembly. Alternatively spliced transcript variants have been identified, but the biological validity of some of these variants has not been determined.
Specifications
| ATPAF1 | |
| Polyclonal | |
| Unconjugated | |
| ATPAF1 | |
| 6330547J17Rik; AI593252; ATP synthase mitochondrial F1 complex assembly factor 1; ATP11; ATP11 homolog; ATP11p; ATPAF1; homolog of yeast ATP11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64756 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5TC12 | |
| ATPAF1 | |
| Recombinant Human ATPAF1. Recombinant protein control fragment (Product #RP-105788). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion