missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPase Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98188JF646
This item is not returnable.
View return policy
Description
ATPase Polyclonal antibody specifically detects ATPase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ATPase | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| ATPase, axonemal, dynein, axonemal, heavy chain 8, dynein, axonemal, heavy polypeptide 8, FLJ25850, FLJ36115, FLJ36334, hdhc9 | |
| Produced in rabbits immunized with E. coli-derived Human ATPase fragment. | |
| 0.1 mL | |
| Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers | |
| 1769 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction