missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPB Antibody (OTI4E5), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70224UV
This item is not returnable.
View return policy
Description
ATPB Monoclonal specifically detects ATPB in Human, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| ATPB | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ATP5F1B | |
| Full length recombinant protein of human ATPB (NP_001677) produced in HEK293T cells. | |
| 0.1 mL | |
| 506.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI4E5 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| ATP synthase subunit beta, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide, ATPMB, ATPSBmitochondrial ATP synthase beta subunit, EC 3.6.3, EC 3.6.3.14, MGC5231, mitochondrial ATP synthetase, beta subunit | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Rat, Canine, Primate, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction