missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ATPG Polyclonal antibody specifically detects ATPG in Human,Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ATPG |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Tris-citrate/phosphate pH 7 -8. |
| Gene Alias | ATP synthase gamma chain, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1, ATP5CATP synthase subunit gamma, mitochondrial, ATP5CL1mitochondrial ATP synthase, gamma subunit 1, F-ATPase gamma subunit |
| Host Species | Rabbit |
| Immunogen | Between 175 and 225 |
| Purification Method | Affinity purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?