Learn More
ATR Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-323AF350
This item is not returnable.
View return policy
Description
ATR Polyclonal antibody specifically detects ATR in Human, Mouse samples. It is validated for Western Blot
Specifications
| ATR | |
| 50mM Sodium Borate | |
| Human |
| Alexa Fluor 350 | |
| RUO | |
| Store at 4C in the dark. |