missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATR Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-323UV
This item is not returnable.
View return policy
Description
ATR Polyclonal antibody specifically detects ATR in Human, Mouse samples. It is validated for Western Blot
Specifications
| ATR | |
| Polyclonal | |
| Western Blot | |
| ataxia telangiectasia and Rad3 related, Ataxia telangiectasia and Rad3-related protein, EC 2.7.11.1, FRAP-related protein 1, FRP1FRAP-related protein-1, MEC1, protein kinase ATR, Rad3 related protein, SCKL1, SCKLMEC1, mitosis entry checkpoint 1, homolog, serine/threonine-protein kinase ATR | |
| The immunogen recognized by this antibody maps to a region between residues 400 and 450 of human Ataxia Telangiectasia and Rad3-related using the numbering given in entry NP_001175.1 (GeneID 545). | |
| 0.1 mL | |
| Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Immunology, p53 Pathway, Virology Bacteria and Parasites | |
| 545 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction