missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATRIP Antibody (OTI1G6), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72270AF532
This item is not returnable.
View return policy
Description
ATRIP Monoclonal specifically detects ATRIP in Human samples. It is validated for Western Blot.
Specifications
| ATRIP | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ATRIP | |
| Recombinant protein fragment corresponding to amino acids 42-309 of human ATRIP (NP_569055) produced in E.coli. | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot | |
| OTI1G6 | |
| Western Blot | |
| AGS1, ATM and Rad3-related-interacting protein, ATR interacting protein, ATR-interacting protein, DKFZp762J2115, FLJ12343, MGC20625, MGC21482, MGC26740 | |
| Mouse | |
| 0.1 mL | |
| 84126 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction