missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATRIP Antibody (OTI1G6), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72270UV
This item is not returnable.
View return policy
Description
ATRIP Monoclonal specifically detects ATRIP in Human samples. It is validated for Western Blot.
Specifications
| ATRIP | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ATRIP | |
| Recombinant protein fragment corresponding to amino acids 42-309 of human ATRIP (NP_569055) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot | |
| OTI1G6 | |
| Western Blot | |
| AGS1, ATM and Rad3-related-interacting protein, ATR interacting protein, ATR-interacting protein, DKFZp762J2115, FLJ12343, MGC20625, MGC21482, MGC26740 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair | |
| 84126.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido