missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATRX Antibody (ATRX/7940), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21065R
This item is not returnable.
View return policy
Description
ATRX Monoclonal antibody specifically detects ATRX in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ATRX | |
| Monoclonal | |
| DyLight 550 | |
| alpha thalassemia/mental retardation syndrome X-linked, alpha thalassemia/mental retardation syndrome X-linked (RAD54 (S. cerevisiae)homolog), alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog, S.cerevisiae), ATP-dependent helicase ATRX, ATR2, DNA dependent ATPase and helicase, EC 3.6.1, EC 3.6.4.12, helicase 2, X-linked, Juberg-Marsidi syndrome, MGC2094, MRXHF1, RAD54 homolog, RAD54L, SFM1, SHS, transcriptional regulator ATRX, XH2RAD54, X-linked helicase II, X-linked nuclear protein, XNPZNF-HX, Zinc finger helicase, Znf-HX | |
| Recombinant full-length human ATRX protein | |
| 0.1 mL | |
| Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair | |
| 546 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| ATRX/7940 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction