missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATRX, Mouse anti-Human, Clone: 5B3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00000546-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: FNLGALSAMSNQQLEDLINQGREKVVEATNSVTAVRIQPLEDIISAVWKENMNLSEAQVQALALSRQASQELDVKRREAIYNDVLTKQQMLISCVQRILMSpecifications
| ATRX | |
| Monoclonal | |
| Unconjugated | |
| alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog, S. cerevisiae) | |
| ATR2/MGC2094/MRXHF1/RAD54/RAD54L/SFM1/SHS/XH2/XNP/ZNF-HX | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 5B3 | |
| In 1x PBS, pH 7.4 | |
| NM_000489 | |
| ATRX | |
| ATRX (NP_000480, 2311 a.a. ∼ 2410 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 546 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction