missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATRX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584664
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: EFRAMDAVNK EKNTKEHKVI DAKFETKARK GEKPCALEKK DISKSEAKLS RKQVDSEHMH QNVPTEEQRT NKSTGGEHKK SDRKEEPQYE PANTSE Highest antigen sequence indentity to the following orthologs: Rat - 57%, Mouse - 51%.
ATRX encodes a protein that contains an ATPase/helicase domain, and thus it belongs to the SWI/SNF family of chromatin remodeling proteins. The mutations of this gene are associated with an X-linked mental retardation (XLMR) syndrome most often accompanied by alpha-thalassemia (ATRX) syndrome. These mutations have been shown to cause diverse changes in the pattern of DNA methylation, which may provide a link between chromatin remodeling, DNA methylation, and gene expression in developmental processes. This protein is found to undergo cell cycle-dependent phosphorylation, which regulates its nuclear matrix and chromatin association, and suggests its involvement in the gene regulation at interphase and chromosomal segregation in mitosis. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.
Spezifikation
| ATRX | |
| Polyclonal | |
| Unconjugated | |
| ATRX | |
| 4833408C14Rik; AI447451; alpha thalassemia/mental retardation syndrome (X-linked); alpha thalassemia/mental retardation syndrome X-linked; alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog, S. cerevisiae); alpha thalassemia/mental retardation syndrome X-linked homolog; alpha thalassemia/mental retardation syndrome X-linked homolog (human); ATP-dependent helicase ATRX; ATR2; ATRX; ATRX, chromatin remodeler; DNA dependent ATPase and helicase; DXHXS6677E; helicase 2; helicase 2, X-linked; helicase II; heterochromatin protein 2; HP1 alpha-interacting protein; Hp1bp2; Hp1bp38; HP1-BP38; HP1-BP38 protein; JMS; mental retardation, X-linked 52; MRX52; MRXHF1; MRXS3; pABP-2; Rad54; RAD54L; RP5-875J14.1; SFM1; SHS; transcriptional regulator ATRX; XH2; X-linked helicase II; X-linked nuclear protein; XNP; Zinc finger helicase; Znf-HX | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 546 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P46100 | |
| ATRX | |
| Recombinant protein corresponding to Human ATRX. Recombinant protein control fragment (Product #RP-107357). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur