missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ Laser Upgrade Kits

Product Code. 17189705
Change view
Click to view available options
Color:
Green
Red
Violet
Yellow
For Use With (Equipment):
Attune Acoustic Focusing Cytometer
Attune CytPix Flow Cytometer
Unit Size:
1 set
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color For Use With (Equipment) unitSize
17189705 Green Attune Acoustic Focusing Cytometer 1 set
15300667 Violet Attune Acoustic Focusing Cytometer Each
15310667 Red Attune Acoustic Focusing Cytometer Each
15320667 Yellow Attune Acoustic Focusing Cytometer Each
17311939 Red Attune CytPix Flow Cytometer Each
17321939 Yellow Attune CytPix Flow Cytometer Each
17331939 Violet Attune CytPix Flow Cytometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 17189705 Supplier Invitrogen™ Supplier No. A32701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enhance the capabilities of your Attune NxT Acoustic Focusing Flow Cytometer and Attune CytPix Imaging Enhanced Flow Cytometer with our versatile laser upgrade kits.

Enhance the capabilities of your Attune NxT Acoustic Focusing Flow Cytometer and Attune CytPix Imaging Enhanced Flow Cytometer with our versatile laser upgrade kits. Available in red, yellow, violet, and green options, these upgrade kits are designed to expand your instrument's fluorescence detection capabilities, enabling a broader range of research applications and enhancing the versatility of your flow cytometry experiments.

Expanded capabilities

Maximize your research potential with our laser upgrade kits. Each kit is tailored to specific excitation wavelengths, allowing you to utilize a variety of labels, dyes, and applications. For example, the Red Laser Upgrade Kit expands the 637-nm excitation capabilities, facilitating the detection of APC, Alexa Fluor 700, APC-tandem dyes, and functional reagents such as CellTrace Far Red dye, LIVE/DEAD Fixable far red or near IR dyes, Vybrant DyeCycle Ruby dye, and far red–exciting fluorescent proteins. Similar enhancements are available with the yellow, violet, and green laser upgrade kits, each broadening the scope of detectable fluorophores and applications.

Cost-effective solutions

Upgrading your existing flow cytometer is a cost-effective way to extend its capabilities without the need to invest in new instruments. These upgrades allow you to configure your Attune NxT and Attune CytPix flow cytometers with different laser configurations, meeting the evolving demands of your research while optimizing your budget.

Simple installation

The Attune NxT and Attune CytPix flow cytometers are designed for easy upgrades. Each laser upgrade kit includes all necessary optical components and can be installed by a qualified service engineer in typically less than four hours. This minimizes downtime and ensures that your instrument is quickly back in operation with enhanced functionality.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Attune
Content And Storage Green laser with optics fiber and optical block
Quantity 1 Kit
Color Green
Format Tube(s)
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Flow Cytometer Laser Lines 532 nm
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Laser Upgrade Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.