Learn More
Attune™ NxT Emission Filter, 780/60BP
Attune NxT Emission Filter, 780/60BP
Brand:  Invitrogen 100022766
Additional Details : Weight : 0.10000kg
Product Code. 15390647
  • € 187.00 / Each
Estimated Shipment: 23-05-2024
to see stock.
Description
Description
This is a replacement 780/60 bandpass filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.
Specifications
Specifications
Attune Acoustic Focusing Cytometer
Room temperature
Attune™ NxT Emission Filter
1 Each