missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atxn1, Mouse anti-Mouse, ATTO488, Clone: S76-8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB18589.100ug
This item is not returnable.
View return policy
Specifications
| Atxn1 | |
| Monoclonal | |
| Atto 488 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
| Ataxin-1/Atx1/C85907/Sca1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| S76-8 | |
| Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| ataxin 1 | |
| Atxn1 | |
| A synthetic peptide corresponding to amino acids 164-197 of mouse Atxn1. | |
| RUO | |
| 20238 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction