missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATXN7L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97598-50ul
This item is not returnable.
View return policy
Description
ATXN7L1 Polyclonal antibody specifically detects ATXN7L1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ATXN7L1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| ataxin 7-like 1, ataxin 7-like 4, ataxin-7-like protein 1, Ataxin-7-like protein 4, ATXN7L4, FLJ40255, FLJ58242, KIAA1218MGC10760, MGC33190 | |
| Produced in rabbits immunized with E. coli-derived Human ATXN7L1 fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 222255 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto