missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATXN7L1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15782341 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15782341 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15782341 Supplier Invitrogen™ Supplier No. PA554412

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SLPLSFDKSE GKKRKNSSSS SKACKITKMP GMNSVHKKNP PSLLAPVPDP VNSTSSRQVG KNSSLALSQS SPSSISSPGH SRQKNTNRTG RIR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 86%.

ATXN7L1 (ataxin-7-like protein 1) is an 833 amino acid protein that contains one SCA7 domain. The ATXN7L1 gene is conserved in chimpanzee, canine, mouse, rat and chicken, and maps to human chromosome 7q22.3. Chromosome 7 is about 158 million bases long, encodes over 1,000 genes and makes up about 5%of the human genome. Chromosome 7 has been linked to osteogenesis imperfecta, Pendred syndrome, lissencephaly, citrullinemia and Shwachman Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterized by mild mental retardation, an unusual comfort and friendliness with strangers and an elfin appearance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATXN7L1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATXN7L1
Gene Accession No. Q9ULK2
Gene Alias 2810423G08Rik; ataxin 7 like 1; ataxin 7-like 1; ataxin 7-like 4; ataxin-7-like protein 1; Ataxin-7-like protein 4; Atxn7l1; Atxn7l4; KIAA1218; RGD1305730
Gene Symbols ATXN7L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATXN7L1. Recombinant protein control fragment (Product #RP-92748).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222255
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.