missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATXN7L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565906
This item is not returnable.
View return policy
Description
Immunogen sequence: KTKTALSMGLN GTMGPRVKRA GPLDCRGSPH QLPTPVKASQ LENRGAAGHP AKALPTNCLS EEEVAKKRKN LATYCRPVKA KHCQAGAPAD V Highest antigen sequence identity to the following orthologs - mouse 87%, rat 86%.
SCA7 is an autosomal dominant neurodegenerative disorder characterized by ataxia and selective neuronal cell loss caused by the expansion of a translated CAG repeat encoding a polyglutamine tract in ataxin-7, which is the SCA7 gene product. Ataxin-7 is a nuclear protein that is expressed within neurons both affected and unaffected in SCA7 pathology with subcellular localization being variable depending upon the neuronal subtype. Polyglutamine expanded in ataxin-7 may carry out its pathogenic effects in the nucleus by altering the matrix-associated nuclear structure and/or by disrupting nucleolar function.ATXN7L2 (Ataxin-7-like protein 2) is a 722 amino acid protein that contains a SCA7 domain, which is highly conserved through all members of the ATXN7 gene family. The gene encoding ATXN7L2 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome.
Specifications
| ATXN7L2 | |
| Polyclonal | |
| Unconjugated | |
| ATXN7L2 | |
| 2610528J18Rik; ataxin 7 like 2; ataxin 7-like 1; ataxin 7-like 2; ataxin-7-like protein 2; ATXN7L2; mFLJ00381 protein; RGD1307047 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 127002 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T6C5 | |
| ATXN7L2 | |
| Recombinant Human ATXN7L2. Recombinant protein control fragment (Product #RP-106585). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction