missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATXN7L3 Monoclonal Antibody (2ATX-2B1)
GREENER_CHOICE

Product Code. 15815873
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15815873 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15815873 Supplier Invitrogen™ Supplier No. MA3084

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA3-084 detects Ataxin 7L3 from human and mouse samples. MA3-084 has been successfully used in Western blot, immunofluorescence, and ELISA applications.

Component of the transcription regulatory histone acetylation (HAT) complex SAGA, a multiprotein complex that activates transcription by remodeling chromatin and mediating histone acetylation and deubiquitination. Within the SAGA complex, participates in a subcomplex that specifically deubiquitinates both histones H2A and H2B (PubMed:18206972, PubMed:21746879). The SAGA complex is recruited to specific gene promoters by activators such as MYC, where it is required for transcription. Required for nuclear receptor-mediated transactivation. Within the complex, it is required to recruit USP22 and ENY2 into the SAGA complex (PubMed:18206972). Regulates H2B monoubiquitination (H2Bub1) levels. Affects subcellular distribution of ENY2, USP22 and ATXN7L3B (PubMed:27601583). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATXN7L3
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2ATX-2B1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene ATXN7L3
Gene Accession No. A2AWT3, Q14CW9
Gene Alias AT7L3; ataxin 7 like 3; ataxin 7-like 3; ataxin-7-like protein 3; ATXN7L3; DKFZp761G2113; E030022H21Rik; SAGA-associated factor 11 homolog
Gene Symbols ATXN7L3
Host Species Mouse
Immunogen Synthetic Peptide coupled to Ovalbumin corresponding to aa 278 to aa 296 of human ataxin 7L3.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217218, 56970
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.