missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: LLWSLFVPFT VYQVRWLRPV HRQLGEANEE FALRVQQLVA KELGQTGTRL TPADKAEHMK RQRHPRLRPQ SAQSSFPPSP GPSPDVQLAT LAQRVKEVLP HVPLGVIQRD LAKTGCVDLT ITNLLEGAVA FMPEDITKGT QSLPT Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
AUP1 contains a domain with homology to the ancient conserved region of the archain 1 gene and a domain they may be involved in binding ubiquitin-conjugating enzymes. The unprocessed precursor is of 476 amino acids in length and has an estimated molecular weight of 53 kDa.
Specifications
Specifications
| Antigen | AUP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | AUP1 |
| Gene Accession No. | A1L134, P70295, Q9Y679 |
| Gene Alias | AA589454; ancient ubiquitous protein 1; ancient ubiquitous protein 1 isoform 2; ancient ubiquitous protein 1; LOW QUALITY PROTEIN: ancient ubiquitous protein 1; Aup1; AUP1 lipid droplet regulating VLDL assembly factor; AUP1, lipid droplet regulating VLDL assembly factor; HUBCEP52; LOW QUALITY PROTEIN: ancient ubiquitous protein 1; RPL40 |
| Gene Symbols | AUP1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
