missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AURKB Mouse anti-Human, Clone: AURKB/1521, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15054.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against partial recombinant human AURKB.
- High specificity
- High reactivity
Specifications
| AURKB | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Q96GD4 | |
| AURKB | |
| Recombinant protein corresponding to amino acids 89-251 of human AURKB. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Western Blot | |
| AURKB/1521 | |
| Mouse monoclonal antibody raised against partial recombinant human AURKB. | |
| In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
| AIK2/AIM-1/AIM1/ARK2/AurB/IPL1/STK12/STK5 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 9212 | |
| Store at 4°C. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction