missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora A/B Antibody (3H1), Novus Biologicals™
Mouse Monoclonal Antibody
€ 220.00 - € 464.00
Specifications
| Antigen | Aurora A/B |
|---|---|
| Clone | 3H1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/ Immunofluorescence 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667927
|
Novus Biologicals
NBP2-50043SS |
0.025 mL |
€ 220.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696368
|
Novus Biologicals
NBP2-50043 |
0.1 mL |
€ 464.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aurora A/B Monoclonal antibody specifically detects Aurora A/B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Aurora A/B | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| O14965, Q96GD4, O14965 | |
| Recombinant full length human Aurora A expressed in and purified from E. coli. [UniProt# O14965] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 3H1 | |
| Western Blot 1:1000, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/ Immunofluorescence 1:500 - 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50% PBS, 50% glycerol | |
| AIK;AIK2;AIM1;AIM-1;AIRK1;AIRK2;ARK1;ARK2;AURA;AurB;AURKA;AURKB;aurkb-sv1;aurkb-sv2;AYK1;BTAK;IAK1;IPL1;PPP1R47;PPP1R48;STK1;STK12;STK15;STK5;STK6;STK7 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title