missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aurora A/B Monoclonal Antibody (3H1)

Mouse Monoclonal Antibody
€ 504.00 - € 1750.00
Specifications
| Antigen | Aurora A/B |
|---|---|
| Clone | 3H1 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17825257
|
Invitrogen™
MA547320 |
500 μL |
€ 1750.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17875277
![]() |
Invitrogen™
MA547418 |
100 μL |
€ 504.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aurora A/B Monoclonal Antibody for Western Blot, ICC/IF, IHC
The Aurora gene, a serine-threonine kinase, orginally discovered in Drosophila, has been found to be responsible for a chromosomal segregation defect. The mammalian homologues of Drosophila Aurora have been termed ARK1 (Aurora-related kinase 1) and ARK2.Specifications
| Aurora A/B | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 5 mM sodium azide | |
| O14965, O55099, O70126, P59241, P97477, Q96GD4 | |
| AURKA, AURKB | |
| Primary | |
| Protein A/G |
| 3H1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human, Mouse, Rat | |
| AURKA | |
| 114592, 20877, 20878, 261730, 6790, 9212 | |
| Full length recombinant human Aurora A protein expressed in and purified from E. coli. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title