missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora C Antibody (OTI2B7), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71303AF405
This item is not returnable.
View return policy
Description
Aurora C Monoclonal specifically detects Aurora C in Human samples. It is validated for Western Blot.
Specifications
| Aurora C | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AURKC | |
| Full-length protein expressed in 293T cell transfected with human Aurora C expression vector. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI2B7 | |
| Western Blot | |
| AIE2AIK3, ARK3Serine/threonine-protein kinase aurora-C, AurC, aurora kinase CAurora/IPL1/Eg2 protein 2, Aurora/IPL1-related kinase 3, aurora-C, Aurora-related kinase 3, EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 13 (aurora/IPL1-like), serine/threonine-protein kinase 13, STK13ARK-3 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 6795.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction